Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11orf73 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989473
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9473 Supplier Bioss Supplier No. BS9942R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for organization and/or function of the secretory apparatus in Clara cells in lung.

Specifications

Antigen C11orf73
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HIKESHI
Gene Accession No. Q53FT3
Gene Alias 0610007P06Rik; 1110002N09Rik; BOS_24379; C11orf73; C21H11orf73; C29H11orf73; C7H11orf73; chromosome 21 open reading frame, human C11orf73; chromosome 29 open reading frame, human C11orf73; chromosome 7 open reading frame, human C11orf73; hikeshi; Hikeshi, heat shock protein nuclear import factor; HLD13; HSPC138; HSPC179; HSPC248; hypothetical protein LOC550239; l(7)6Rn; L7rn6; Lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; LOC100343781; OPI10; Protein Hikeshi; uncharacterized protein C11orf73 homolog; zgc:110091
Gene Symbols HIKESHI
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C11orf73.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51501
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.