Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf79 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558796 Supplier Invitrogen™ Supplier No. PA558796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAKEHLQHMT EKQLNLYDRL INEPSNDWDI YYWATEAKPA PEIFENEVMA LLRDFAKNKN KEQRLRAPDL EYLFE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

SDHAF2, also named asC11orf79, SDHAF2 and SDH5, is a mitochondrial protein needed for the flavination of a succinate dehydrogenase complex subunit required for activity of the complex. SDHAF2 has been shown to be mutated in patients with paragangliomas, also known as familial non-chromaffin paragangliomas type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf79
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sdhaf2
Gene Accession No. Q9NX18
Gene Alias 0610038F07Rik; AA407634; AW049997; C11orf79; HAMAP-Rule:MF_03057}; hSDH5; PGL2; RGD1309216; SDH assembly factor 2; SDH assembly factor 2 {ECO:0000255; SDH5; SDHAF2; succinate dehydrogenase assembly factor 2, mitochondrial; succinate dehydrogenase assembly factor 2, mitochondrial {ECO:0000255; succinate dehydrogenase complex assembly factor 2; succinate dehydrogenase subunit 5, mitochondrial; succinate dehydrogenase subunit 5, mitochondrial {ECO:0000255
Gene Symbols Sdhaf2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C11orf79. Recombinant protein control fragment (Product #RP-97444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.