Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C11orf79 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595718 Supplier Invitrogen™ Supplier No. PA595718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVSTVFSTS SLMLALSRHS LLSPLLSVTS FRRFYRGDSP TDSQKDMIEI PLPPWQERTD ESIETKRARL LYESRKRGML ENCILLSLFA KEHLQHMTEK QLNLYDRLIN EPSNDWDIYY WATEAKPAPE IFENEVMALL RDFAKNKNKE QRLRAPDLEY LFEKPR; Positive Samples: HeLa, mouse kidney, mouse stomach, mouse brain.

SDHAF2, also named asC11orf79, SDHAF2 and SDH5, is a mitochondrial protein needed for the flavination of a succinate dehydrogenase complex subunit required for activity of the complex. SDHAF2 has been shown to be mutated in patients with paragangliomas, also known as familial non-chromaffin paragangliomas type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf79
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Sdhaf2
Gene Accession No. Q8C6I2, Q9NX18
Gene Alias 0610038F07Rik; AA407634; AW049997; C11orf79; HAMAP-Rule:MF_03057}; hSDH5; PGL2; RGD1309216; SDH assembly factor 2; SDH assembly factor 2 {ECO:0000255; SDH5; SDHAF2; succinate dehydrogenase assembly factor 2, mitochondrial; succinate dehydrogenase assembly factor 2, mitochondrial {ECO:0000255; succinate dehydrogenase complex assembly factor 2; succinate dehydrogenase subunit 5, mitochondrial; succinate dehydrogenase subunit 5, mitochondrial {ECO:0000255
Gene Symbols Sdhaf2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-166 of human SDHAF2 (NP_0603111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54949, 66072
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.