Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568366 Supplier Invitrogen™ Supplier No. PA568366
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C12orf10 Polyclonal Antibody for IHC (P), Western Blot

C12orf10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene C12orf10
Gene Accession No. Q9HB07
Gene Alias C12orf10; chromosome 12 open reading frame 10; Gamm1; melanocyte proliferating gene 1; melanocyte related; MST024; MSTP024; MYG; MYG1; UPF0160 protein MYG1, mitochondrial
Gene Symbols C12orf10
Host Species Rabbit
Immunogen Synthetic protein corresponding to residues near the C terminal of human C12orf10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60314
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.