Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558473 Supplier Invitrogen™ Supplier No. PA558473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYALTTTLSA RVARLNPTWN HPDQDTEAGF KRAMDLVQEE FLQRLDFYQH SWLPARALVE EALAQRFQVD P Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

C12orf10 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C12orf10
Gene Accession No. Q9HB07
Gene Alias C12orf10; chromosome 12 open reading frame 10; Gamm1; melanocyte proliferating gene 1; melanocyte related; MST024; MSTP024; MYG; MYG1; UPF0160 protein MYG1, mitochondrial
Gene Symbols C12orf10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C12orf10. Recombinant protein control fragment (Product #RP-102842).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60314
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.