Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598886
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598886 Supplier Invitrogen™ Supplier No. PA598886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C12orf11 Polyclonal Antibody for Western Blot, IP, ELISA

Crucial regulator of the mitotic cell cycle and development. At prophase, required for dynein anchoring to the nuclear envelope important for proper centrosome-nucleus coupling. At G2/M phase, may be required for proper spindle formation and execution of cytokinesis. Probable component of the Integrator (INT) complex, a complex involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing (PubMed:23904267). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf11
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene INTS13
Gene Accession No. Q9NVM9
Gene Alias 4933424B01Rik; ASUN; asunder spermatogenesis regulator; asunder, spermatogenesis regulator; asunder, spermatogenesis regulator homolog (Drosphila); C12orf11; C27H12orf11; C5H12orf11; Cell cycle regulator Mat89Bb homolog; Cell cycle regulator Mat89Bb-like protein-like protein; fi39e09; GCT1; Germ cell tumor 1; integrator complex subunit 13; integrator complex subunit 13; protein asunder homolog; IntS13; likely orthologue of H. sapiens chromosome 12 open reading frame 11 (C12orf11); Mat89Bb; NET48; Protein asunder homolog; sarcoma antigen NY-SAR-95; Similar to hypothetical protein FLJ10637; similar to Protein C12orf11 (Sarcoma antigen NY-SAR-95); SPATA30; spermatogenesis associated 30; spermatogenesis-associated protein 30; wu:fi39e09; zgc:55466
Gene Symbols INTS13
Host Species Rabbit
Immunogen Recombinant Human Integrator complex subunit 13 protein (573-706AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55726
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.