Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C12orf11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114884
Description
Antibody detects endogenous levels of total ASUN.
Crucial regulator of the mitotic cell cycle and development. At prophase, required for dynein anchoring to the nuclear envelope important for proper centrosome-nucleus coupling. At G2/M phase, may be required for proper spindle formation and execution of cytokinesis. Probable component of the Integrator (INT) complex, a complex involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing (PubMed:23904267). [UniProt].
Specifications
| C12orf11 | |
| Polyclonal | |
| Unconjugated | |
| INTS13 | |
| 4933424B01Rik; ASUN; asunder spermatogenesis regulator; asunder, spermatogenesis regulator; asunder, spermatogenesis regulator homolog (Drosphila); C12orf11; C27H12orf11; C5H12orf11; Cell cycle regulator Mat89Bb homolog; Cell cycle regulator Mat89Bb-like protein-like protein; fi39e09; GCT1; Germ cell tumor 1; integrator complex subunit 13; integrator complex subunit 13; protein asunder homolog; IntS13; likely orthologue of H. sapiens chromosome 12 open reading frame 11 (C12orf11); Mat89Bb; NET48; Protein asunder homolog; sarcoma antigen NY-SAR-95; Similar to hypothetical protein FLJ10637; similar to Protein C12orf11 (Sarcoma antigen NY-SAR-95); SPATA30; spermatogenesis associated 30; spermatogenesis-associated protein 30; wu:fi39e09; zgc:55466 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55726, 690728, 71177 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8QZV7, Q9NVM9 | |
| INTS13 | |
| A synthesized peptide derived from human INTS13(Accession Q9NVM9), corresponding to amino acid residues P642-E692. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction