Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544240 Supplier Invitrogen™ Supplier No. PA544240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AVAGTEGGGG GSAGYSCYQN SKGSDRIKDG YKVNSHIAKL QELWKTPQNQ Sequence homology: Cow: 79%; Human: 100%; Pig: 79%.

The exact function of C12orf24 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf24
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM216A
Gene Accession No. Q8WUB2
Gene Alias 1500011H22Rik; C12orf24; C14H12orf24; C17H12orf24; FAM216A; family with sequence similarity 216 member A; family with sequence similarity 216, member A; HSU79274; hypothetical protein LOC616613; protein FAM216A; protein predicted by clone 23733; RGD1310861; uncharacterized protein C12orf24; uncharacterized protein C12orf24 homolog
Gene Symbols FAM216A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C12orf24 (aa 38-87).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29902
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.