Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C12ORF24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989476
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9476 Supplier Bioss Supplier No. BS9945R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The exact function of C12orf24 remains unknown.

Specifications

Antigen C12ORF24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM216A
Gene Accession No. Q8WUB2
Gene Alias BOS_16570; C12orf24; C14H12orf24; C17H12orf24; chromosome 12 open reading frame 24 ortholog; chromosome 17 open reading frame, human C12orf24; FAM216A; family with sequence similarity 216 member A; family with sequence similarity 216, member A; HSU79274; Protein FAM216A; protein predicted by clone 23733; uncharacterized protein C12orf24; uncharacterized protein C12orf24 homolog
Gene Symbols FAM216A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C12ORF24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29902
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.