Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558873 Supplier Invitrogen™ Supplier No. PA558873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QIRNLPSLKH NDLLSWFEDC KEGKIEGIVW HCSDGCLIKV HRHHLGLCWP IPDTYMNSRP VIINMNLNKC DSA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

C12orf29 (Chromosome 12 Open Reading Frame 29) is a Protein Coding gene. Diseases associated with C12orf29 include Senior-Loken Syndrome 6 and Meckel Syndrome, Type 4. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf29
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C12orf29
Gene Accession No. Q8N999
Gene Alias C12orf29; chromosome 12 open reading frame 29; CLO29; DKFZp313K0436; DKFZp434N2030; DKFZp686L04169; FLJ38158; MGC102978; uncharacterized protein C12orf29
Gene Symbols C12orf29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C12orf29. Recombinant protein control fragment (Product #RP-97732).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.