Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537983
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537983 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537983 Supplier Invitrogen™ Supplier No. PA537983
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with pig, sheep and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

C12orf29 (Chromosome 12 Open Reading Frame 29) is a Protein Coding gene. Diseases associated with C12orf29 include Senior-Loken Syndrome 6 and Meckel Syndrome, Type 4. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf29
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene C12orf29
Gene Accession No. Q8N999
Gene Alias C12orf29; chromosome 12 open reading frame 29; CLO29; DKFZp313K0436; DKFZp434N2030; DKFZp686L04169; FLJ38158; MGC102978; uncharacterized protein C12orf29
Gene Symbols C12orf29
Host Species Goat
Immunogen Peptide with sequence C-QPFKVLATETVSHK, from the internal region of C12orf29 (aa30-43).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91298
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.