Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597099 Supplier Invitrogen™ Supplier No. PA597099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNHPVEPKNS EKTAENGVSS RIDILRLLLQ QLEATLETGK RFIEKDIQYP FLGPVPTRMG GFFNSGCSQC QISSFYLVND IYELDTSGLE DTMEIQERIE NSFKSLLDQL KDVFSKCKGD LLEVKDGNLK THPTLLENLV FFVETISVIL WVSSYCESVL RPYKLNLQKK KKKKKETSII MPPVFTSFQD YVTGLQTLIS NVVDHIKGLE THLIALKLEE LILEDTSLSP EERKFSKTVQ GKVQSSYLHS LLEMGELLKK RLETTKKLKI; Positive Samples: Mouse uterus, Mouse eye, Mouse ovary, Rat uterus, Rat eye, Rat ovary; Cellular Location: Cytoplasm.

This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf30
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Naa25
Gene Accession No. Q6QI44, Q8BWZ3
Gene Alias 4833422K13Rik; A630046C11; AI450868; C12orf30; C330023M02Rik; Liver regeneration-related protein LRRGT00164; LRRGT00164; MDM20; mitochondrial distribution and morphology 20; mitochondrial distribution and morphology protein 20; N(alpha)-acetyltransferase 25, NatB auxiliary subunit; Naa25; Nalpha acteyltransferase 25; N-alpha-acetyltransferase 25, NatB auxiliary subunit; NAP1; NatB complex subunit MDM20; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; p120; RGD1305685
Gene Symbols Naa25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 703-972 of human NAA25 (NP_0792292).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231713, 360811
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.