Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C12orf30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598409
Description
C12orf30 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.
Specifications
C12orf30 | |
Polyclonal | |
Unconjugated | |
Naa25 | |
4833422K13Rik; A630046C11; AI450868; C12orf30; C330023M02Rik; Liver regeneration-related protein LRRGT00164; LRRGT00164; MDM20; mitochondrial distribution and morphology 20; mitochondrial distribution and morphology protein 20; N(alpha)-acetyltransferase 25, NatB auxiliary subunit; Naa25; Nalpha acteyltransferase 25; N-alpha-acetyltransferase 25, NatB auxiliary subunit; NAP1; NatB complex subunit MDM20; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; p120; RGD1305685 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
231713, 80018 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14CX7, Q8BWZ3 | |
Naa25 | |
Recombinant Human N-alpha-acetyltransferase 25, NatB auxiliary subunit protein (703-972AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction