Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C12orf32 Mouse anti-Human, Clone: OTI2C12, lyophilized, TrueMAB™
SDP

Catalog No. 501670588
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0588 Supplier OriGene Supplier No. CF811314
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

RHNO1 plays a role in DNA damage response (DDR) signaling upon genotoxic stresses such as ionizing radiation (IR) during the S phase. Recruited to sites of DNA damage through interaction with the 9-1-1 cell-cycle checkpoint response complex and TOPBP1 in a ATR-dependent manner. Required for the progression of the G1 to S phase transition. Plays a role in the stimulation of CHEK1 phosphorylation.

Specifications

Antigen C12orf32
Applications Western Blot
Classification Monoclonal
Clone OTI2C12
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene RHNO1
Gene Accession No. Q9BSD3
Gene Alias C12orf32, HKMT1188, RHINO
Gene Symbols RHNO1
Host Species Mouse
Immunogen Full length human recombit protein of human C12ORF32 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83695
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.