Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C12orf4 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321423JF549
Description
C12orf4 Polyclonal antibody specifically detects C12orf4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
C12orf4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
chromosome 12 open reading frame 4, hypothetical protein LOC57102 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C12orf4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
57102 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction