Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112104 Supplier Invitrogen™ Supplier No. PA5112104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: Human C12orf4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene C12orf4
Gene Accession No. Q9NQ89
Gene Alias C12orf4; chromosome 12 open reading frame 4; protein C12orf4
Gene Symbols C12orf4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human C12orf4.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57102
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.