Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558416 Supplier Invitrogen™ Supplier No. PA558416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSDDLDVVGD GMQCPPSPLL FDPSLTLEDH LVKEIAEDPV DILGQMQMAG DGCRSQGSRN SEKASAPLSQ SGLATANGKP EP Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.

The C12orf41 gene, also known as CFAP70 (Cilia and Flagella Associated Protein 70), is located on chromosome 12 and encodes a protein that is essential for the proper function of cilia and flagella. The CFAP70 protein plays a critical role in the assembly and maintenance of axonemal structures, which are core components of cilia and flagella. Cilia and flagella are vital for various cellular processes, including motility, fluid movement across cell surfaces, and signal transduction. Mutations in CFAP70 have been linked to primary ciliary dyskinesia (PCD), a genetic disorder characterized by chronic respiratory tract infections, abnormal situs (organ positioning), and infertility due to defective motility of cilia and flagella. CFAP70 is also involved in the regulation of dynein arms and radial spokes, which are necessary for the effective beating of cilia and flagella. Research into CFAP70 continues to uncover its multifaceted roles in cellular function and its potential implications in ciliopathies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf41
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KANSL2
Gene Accession No. Q9H9L4
Gene Alias 2310037I24Rik; B930009D17; C12orf41; Kansl2; KAT8 regulatory NSL complex subunit 2; non-specific lethal 2 homolog; NSL complex protein NSL2; NSL2
Gene Symbols KANSL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C12orf41. Recombinant protein control fragment (Product #RP-97768).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54934
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.