Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C12orf50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | C12orf50 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
C12orf50 Polyclonal specifically detects C12orf50 in Human samples. It is validated for Western Blot.Specifications
| C12orf50 | |
| Polyclonal | |
| Rabbit | |
| Q8NA57 | |
| 160419 | |
| Synthetic peptides corresponding to C12ORF50 The peptide sequence was selected from the N terminal of C12ORF50. Peptide sequence INGLFLPPSSNITLQKEIQEGIPLQSQSQEPLKPQENISRPIHHPLVLKT. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| chromosome 12 open reading frame 50, FLJ35821, hypothetical protein LOC160419 | |
| C12ORF50 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title