Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552715 Supplier Invitrogen™ Supplier No. PA552715
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRSQKRRRPS GQQGALRQEE SQQPLTDLSP AGVTVLGAFG DSPTPTPDHE EPRGGPRPGM PHPKGAPAFQ LNR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The function of this protein has not been specifically defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf53
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIANP
Gene Accession No. Q8IYJ0
Gene Alias AI255183; AU067746; Brain protein 1; C12orf53; C530028O21Rik; LEDA1; leda-1; liver endothelial differentiation-associated protein-1; paired immunoglobin-like type 2 receptor-associating neural protein; PANP; Pianp; PILR alpha associated neural protein; PILR alpha-associated neural protein; PILR-associating neural protein; RGD1304952; UNQ828/PRO1755
Gene Symbols PIANP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C12orf53. Recombinant protein control fragment (Product #RP-90293).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196500
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.