Learn More
Invitrogen™ C12orf53 Polyclonal Antibody

Description
This antibody is predicted to react with non-human primate and rat based on sequence homology.
Specifications
Specifications
| Antigen | C12orf53 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PIANP |
| Gene Accession No. | Q6P1B3, Q8IYJ0 |
| Gene Alias | AI255183; AU067746; Brain protein 1; C12orf53; C530028O21Rik; LEDA1; leda-1; liver endothelial differentiation-associated protein-1; paired immunoglobin-like type 2 receptor-associating neural protein; PANP; Pianp; PILR alpha associated neural protein; PILR alpha-associated neural protein; PILR-associating neural protein; RGD1304952; UNQ828/PRO1755 |
| Gene Symbols | PIANP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.