Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C12orf54 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140329
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MAQHPCQDQE QKVEMTSKQQ RSTSIEETMR PQEKQVTITE TLWDQVLTVF KDIQKELQED ARIR.

Specifications
C12orf54 | |
Polyclonal | |
Unconjugated | |
C12orf54 | |
C12orf54; chromosome 12 open reading frame 54; HSD29; HSD-29; HSD30; HSD-30; Uncharacterized protein C12orf54 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
121273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6X4T0 | |
C12orf54 | |
Recombinant protein corresponding to Human C12orf54. Recombinant protein control fragment (Product #RP-109316). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction