Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C12orf56 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546114 Supplier Invitrogen™ Supplier No. PA546114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RDVVAIDLID DYPEFLSSPD REISQHIRII YSSTVLKKEC KKSNSVRKFL Sequence homology: Horse: 79%; Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf56
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C12orf56
Gene Accession No. Q8IXR9
Gene Alias C12orf56; chromosome 12 open reading frame 56; uncharacterized protein C12orf56
Gene Symbols C12orf56
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C12orf56 (aa 77-126).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115749
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.