Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C12orf57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565428
Description
Immunogen sequence: MASASTQPAAL SAEQAKVVLA EVIQAFSAPE NAVRMDEARD NACNDMGKML QFVLPVATQI QQ Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
In brain, may be required for corpus callusum development.
Specifications
C12orf57 | |
Polyclonal | |
Unconjugated | |
C12orf57 | |
C10; C12orf57; chromosome 12 open reading frame 57; GRCC10; likely ortholog of mouse gene rich cluster, C10; protein C10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113246 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99622 | |
C12orf57 | |
Recombinant Human C12orf57. Recombinant protein control fragment (Product #RP-97711). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction