Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C12orf74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571021
Description
This target displays homology in the following species: Horse: 100%; Human: 100%; Rabbit: 86%.
C12orf74 is a protein coding gene.
Specifications
C12orf74 | |
Polyclonal | |
Unconjugated | |
PLEKHG7 | |
C12orf74; chromosome 12 open reading frame 74; uncharacterized protein C12orf74 | |
Rabbit | |
Affinity chromatography | |
RUO | |
440107 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZR37 | |
PLEKHG7 | |
synthetic peptide directed towards the N-terminal region of human C12orf74. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction