Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C13orf15 Monoclonal Antibody (3B9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524460
Description
Peptide Sequence: MKPPAEDLSD ALCEFDAVLA DFASPFHERH FHYEEHLERM KRRSSASVSD SSGFSDSESA DSLYRNSFSF SDEKLNSPTD STPALLSATV TPQKAKLGDT KELEAFIADL DKTLASM.
Modulates the activity of cell cycle-specific kinases. Enhances CDK1 activity. May contribute to the regulation of the cell cycle. May inhibit growth of glioma cells by promoting arrest of mitotic progression at the G2/M transition. Fibrogenic factor contributing to the pathogenesis of renal fibrosis through fibroblast activation.
Specifications
C13orf15 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9H4X1 | |
RGCC | |
C13orf15 (NP_054778.1, 1 a.a. ~ 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry | |
3B9 | |
Unconjugated | |
RGCC | |
1190002H23Rik; bA157L14.2; C13orf15; regulator of cell cycle; regulator of cell cycle RGCC; Response gene to complement 32 protein; Rgc32; RGC-32; Rgcc | |
Mouse | |
Protein A | |
RUO | |
28984 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction