Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C13orf27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562802 Supplier Invitrogen™ Supplier No. PA562802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGGRSMGSRA AASVMCHIEP DDGDDFVRGL ICISYPLHHP KQQHKLRDED LFRLKEPVLF VSGSADEMCE KNLLEKVAQK MQAP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

C13orf27 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C13orf27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEX30
Gene Accession No. Q5JUR7
Gene Alias 1700029F09Rik; 3110030D08Rik; C13orf27; testis expressed 30; Testis-expressed protein 30; testis-expressed sequence 30 protein; TEX30
Gene Symbols TEX30
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C13orf27. Recombinant protein control fragment (Product #RP-97977).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.