Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C13orf28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583909 Supplier Invitrogen™ Supplier No. PA583909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGIDENYQAG GSENYHELLE NLQFSPGIED KISNDEANAN ANLHGDPSEN YRGPQVSPGS EKSVSSKEKN SKNTQYENLS ILDQIL Highest antigen sequence indentity to the following orthologs: Rat - 36%, Mouse - 43%.

SPACA7 is a protein coding gene involved in fertilization. Seems not to play a direct role in sperm-egg binding or gamete fusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C13orf28
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPACA7
Gene Accession No. Q96KW9
Gene Alias C13orf28; protein SPACA7; SPACA7; sperm acrosome associated 7; sperm acrosome-associated protein 7
Gene Symbols SPACA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPACA7. Recombinant protein control fragment (Product #RP-108202).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.