Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C13orf7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557059 Supplier Invitrogen™ Supplier No. PA557059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKGSLTNDQL ENGSEWKPTS FFLLSPSDQE MNEDFSLHSS SCPVTNEIKP PSCLFQTEFS QGILLSSSHR LFEDQRFGSS LFK Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 70%.

RNF219 (Ring Finger Protein 219, C13orf7) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C13orf7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF219
Gene Accession No. Q5W0B1
Gene Alias 2610206B13Rik; 2810449K13Rik; AI451544; C13orf7; FLJ13449; H12C13ORF7; Obi1; ORC ubiquitin ligase 1; RGD1564706; RING finger protein 219; RING finger protein 219 pseudogene; RNF219
Gene Symbols RNF219
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C13orf7. Recombinant protein control fragment (Product #RP-94994).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.