Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C13orf7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112698
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112698 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112698 Supplier Invitrogen™ Supplier No. PA5112698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C13orf7 Polyclonal Antibody for Western Blot, IHC (P), ELISA

RNF219 (Ring Finger Protein 219, C13orf7) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C13orf7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RNF219
Gene Accession No. Q5W0B1
Gene Alias 2610206B13Rik; 2810449K13Rik; AI451544; C13orf7; FLJ13449; H12C13ORF7; Obi1; ORC ubiquitin ligase 1; RGD1564706; RING finger protein 219; RING finger protein 219 pseudogene; RNF219
Gene Symbols RNF219
Host Species Rabbit
Immunogen Recombinant Human RING finger protein 219 protein (257-465AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79596
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.