Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C13orf7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542338 Supplier Invitrogen™ Supplier No. PA542338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LVTDNPSKIN PETVAEWKKK LRTANEIYEK VKDDVDKLKE ANKKLKLENG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

RNF219 (Ring Finger Protein 219, C13orf7) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C13orf7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF219
Gene Accession No. Q5W0B1
Gene Alias 2610206B13Rik; 2810449K13Rik; AI451544; C13orf7; FLJ13449; H12C13ORF7; Obi1; ORC ubiquitin ligase 1; RGD1564706; RING finger protein 219; RING finger protein 219 pseudogene; RNF219
Gene Symbols RNF219
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C13orf7 (aa 144-193).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79596
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.