Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C14orf1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560268
Description
Immunogen sequence: QSFRDHTFLY EKLYTGKPNL VNGLQARTF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
C14orf1 is a protein coding gene.
Specifications
C14orf1 | |
Polyclonal | |
Unconjugated | |
ERG28 | |
AD-011; C14orf1; chromosome 14 open reading frame 1; ERG28; HSPC288; NET51; Probable ergosterol biosynthetic protein 28; x0006 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKR5 | |
ERG28 | |
Recombinant protein corresponding to Human C14orf1. Recombinant protein control fragment (Product #RP-100139). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction