Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf1 Polyclonal Antibody

Catalog No. PIPA566599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566599 Supplier Invitrogen™ Supplier No. PA566599

Rabbit Polyclonal Antibody

Immunogen sequence: MLVGLRYLEVE PVSRQKKRN Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

C14orf1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERG28
Gene Accession No. Q9UKR5
Gene Alias AD-011; C14orf1; chromosome 14 open reading frame 1; ERG28; HSPC288; NET51; Probable ergosterol biosynthetic protein 28; x0006
Gene Symbols ERG28
Host Species Rabbit
Immunogen Recombinant Human C14orf1. Recombinant protein control fragment (Product #RP-107253).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.