Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf100 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567190 Supplier Invitrogen™ Supplier No. PA567190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TFRRPMAVDIQ PACLGLYCGK TLLFKNGSTE IYGECGVCPR GQRTNAQKYC QPCTESPELY D Highest antigen sequence identity to the following orthologs - mouse 90%, rat 90%.

May be a regulator of the duration of MAPK8 activity in response to various stress stimuli. Facilitates degradation of misfolded endoplasmic reticulum (ER) luminal proteins through the recruitment of components of the proteasome and endoplasmic reticulum-associated degradation (ERAD) system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf100
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JKAMP
Gene Accession No. Q9P055
Gene Alias C14orf100; CDA06; HSPC213; HSPC327; JAMP; JKAMP; JNK1/MAPK8-associated membrane protein; JNK1-associated membrane protein; JNK-associated membrane protein; Jun N-terminal kinase 1-associated membrane protein; medulloblastoma antigen MU-MB-50.4
Gene Symbols JKAMP
Host Species Rabbit
Immunogen Recombinant Human C14orf100. Recombinant protein control fragment (Product #RP-107830).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.