Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf106 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989201
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9201 Supplier Bioss Supplier No. BS9615R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MIS18BP1 is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.

Specifications

Antigen C14orf106
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MIS18BP1
Gene Accession No. Q6P0N0
Gene Alias C14orf106; HSA242977; HsKNL-2; KIAA1903; kinetochore null 2 homolog; Kinetochore-associated protein KNL-2 homolog; KNL2; M18BP1; MIS18 binding protein 1; Mis18-binding protein 1; MIS18BP1; P243; putative protein p243 which interacts with transcription factor Sp1
Gene Symbols MIS18BP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KNL2/C14orf106.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55320
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.