Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C14orf119 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544687
Description
Peptide sequence: SSSMPLSFPS LLPSVPHNTN PSPPLMSYIT SQEMKCILHW FANWSGPQRE Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
C14orf119 | |
Polyclonal | |
Unconjugated | |
C14orf119 | |
C14orf119; chromosome 14 open reading frame 119; My028; My028 protein; uncharacterized protein C14orf119 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55017 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NWQ9 | |
C14orf119 | |
Synthetic peptide directed towards the N-terminal of human C14orf119 (aa 6-55). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction