Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf126 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551516 Supplier Invitrogen™ Supplier No. PA551516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVEVQRGLVI YVCFFKGADK ELLPKMVNTL LNVKLSETEN GKHVSILDLP GNILIIPQAT LGGRLKGRNM QYHSNSGKEE GFELYSQFVT LCEKEVAANS KCAEARVVVE HGTYGNRQVL KLDTNGPFTH LIFE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Deacylates mischarged D-aminoacyl-tRNAs. Also deacylates mischarged glycyl-tRNA(Ala), protecting cells against glycine mischarging by AlaRS. Probably acts by rejecting L-amino acids from its binding site rather than specific recognition of D-amino acids. Catalyzes the hydrolysis of D-tyrosyl-tRNA(Tyr), has no activity on correctly charged L-tyrosyl-tRNA(Tyr). By recycling D-aminoacyl-tRNA to D-amino acids and free tRNA molecules, this enzyme counteracts the toxicity associated with the formation of D-aminoacyl-tRNA entities in vivo and helps enforce protein L-homochirality. In contrast to DTD1, deacylates L-Ala mischarged on tRNA(Thr)(G4.U69) by alanine-tRNA ligase AARS (PubMed:29410408). Can deacylate L-Ala due to a relaxed specificity for substrate chirality caused by the trans conformation of the Gly-Pro motif in the active site (PubMed:29410408). Also hydrolyzes correctly charged, achiral, glycyl-tRNA(Gly) in vitro, although in vivo EEF1A1/EF-Tu may protect cognate achiral glycyl-tRNA(Gly) from DTD2-mediated deacetylation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf126
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DTD2
Gene Accession No. Q96FN9
Gene Alias 4930578F06Rik; 6530401N04Rik; Animalia-specific tRNA deacylase; ATD; B830049N13Rik; C14orf126; D-aminoacyl-tRNA deacylase 2; DTD2; D-tyrosyl-tRNA deacylase 2; D-tyrosyl-tRNA deacylase 2 (putative); D-tyrosyl-tRNA(Tyr) deacylase 2; L-alanyl-tRNA deacylase; probable D-tyrosyl-tRNA(Tyr) deacylase 2
Gene Symbols DTD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C14orf126. Recombinant protein control fragment (Product #RP-88856).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.