Learn More
Invitrogen™ C14orf129 Monoclonal Antibody (2F2)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524550
Catalog No. PIMA524550
Only null left
Add to Cart
Description
Description
Peptide Sequence: METDCNPMEL SSMSGFEEGS ELNGFEGTDM KDMRLEAEAV VNDVLFAVNN MFVSKSLRCA DDVAYINVET KERNRYCLEL TEAGLKVVGY AFDQVDDHLQ TPYHETVYSL LDTLSPAYRE AFGNALLQRL EALKRDGQS.
This gene encodes a protein that is involved as a negative regulator of GSK3-beta in the Wnt signaling pathway. The encoded protein may play a role in the retinoic acid signaling pathway by regulating the functional interactions between GSK3-beta, beta-catenin and cyclin D1, and it regulates the beta-catenin/N-cadherin pool. The encoded protein contains a GSK3-beta interacting domain (GID) in its C-terminus, which is similar to the GID of Axin. The protein also contains an evolutionarily conserved RII-binding domain, which facilitates binding with protein kinase-A and GSK3-beta, enabling its role as an A-kinase anchoring protein. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C14orf129
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
Q9P0R6
GSKIP
C14orf129 (NP_057556.2, 1 a.a. ~ 139 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot, Immunocytochemistry
2F2
Unconjugated
GSKIP
4933433P14Rik; C14orf129; GSK3B interacting protein; GSK3beta interaction protein; GSK3-beta interaction protein; GSK3B-interacting protein; GSKIP; HSPC210; MGC4945; RGD1308470
Mouse
Protein A
RUO
51527
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.