Learn More
Invitrogen™ C14orf130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113775
Description
Immunogen sequence: MAGAEGAAGR QSELEPVVSL VDVLEEDEEL ENEACAVLGG SDSEKCSYSQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 82%; Zebrafish: 100%.
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway, recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
| C14orf130 | |
| Polyclonal | |
| Unconjugated | |
| UBR7 | |
| 5730410I19Rik; AA589405; AW557761; C14orf130; C21H14ORF130; fb74a06; hypothetical protein LOC322815; LOW QUALITY PROTEIN: putative E3 ubiquitin-protein ligase UBR7; N-recognin-7; Putative E3 ubiquitin-protein ligase UBR7; RGD1359144; RING-type E3 ubiquitin transferase UBR7; ubiquitin protein ligase E3 component n-recognin 7; ubiquitin protein ligase E3 component n-recognin 7 (putative); Ubr7; wu:fb74a06; wu:fi29b06; zgc:55404 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55148 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N806 | |
| UBR7 | |
| Synthetic peptide directed towards the N terminal region of human C14orf130 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.