Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf142 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570950 Supplier Invitrogen™ Supplier No. PA570950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 91%; Pig: 79%; Rat: 91%.

Component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t(6)A37) in tRNAs that read codons beginning with adenine. The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37. GON7 likely plays a supporting role to the catalytic subunit OSGEP in the complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf142
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GON7
Gene Accession No. Q9BXV9
Gene Alias AK010878; C14orf142; C21H14orf142; C24H14orf142; C7H14orf142; C8H14orf142; cDNA sequence AK010878; EKC/KEOPS complex subunit GON7; GON7; GON7 subunit of KEOPS complex; GON7, KEOPS complex subunit homolog; hypothetical protein LOC100233176; LOC100233176; PNAS-127; uncharacterized protein C14orf142; uncharacterized protein C14orf142 homolog
Gene Symbols GON7
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human C14orf142.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84520
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.