Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf145 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551515
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551515 Supplier Invitrogen™ Supplier No. PA551515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEMGSLQDRV IALETSTQVA LDHLESVPEK LSLLEDFKDF RDSCSSSERT DGRYSKYRVR RNSLQHHQDD TKYRTKSFKG DRTFLEGSHT RGLDHSSSWQ DHSRFLSSPR FSYVNSFTKR TVAPDSASNK EDATMNGTSS QPK Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 25%.

CEP128 gene ontology annotations related to this gene include protein localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf145
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEP128
Gene Accession No. Q6ZU80
Gene Alias 4930534B04Rik; 5430424K18Rik; AI449708; AI480494; C14orf145; C14orf61; centrosomal protein 128; centrosomal protein 128kDa; centrosomal protein of 128 kDa; Cep128; E230024F20; LEDP/132; lens epithelium-derived protein; LOC500700; similar to chromosome 14 open reading frame 145
Gene Symbols CEP128
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C14orf145. Recombinant protein control fragment (Product #RP-88854).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145508
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.