Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf159 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559566 Supplier Invitrogen™ Supplier No. PA559566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGAYKTTVPC VTHAGFCCPL VVTMRPIPKD KLEGLVRACC SLGGEQGQPV HMGDPELLGI KELSKPAYGD AMVCPPGEVP VFWPSPLTSL GAVSSCETPL A Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.

C14orf159 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf159
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DGLUCY
Gene Accession No. Q7Z3D6
Gene Alias C14orf159; chromosome 14 open reading frame 159; DGLUCY; D-glutamate cyclase, mitochondrial; UNQ2439/PRO5000; UPF0317 protein C14orf159, mitochondrial
Gene Symbols DGLUCY
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C14orf159. Recombinant protein control fragment (Product #RP-98444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.