Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RTRAF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558944
Description
Immunogen sequence: ALDYHNPAGF NCKDETEFRN FIVWLEDQKI RHYKIEDRGN LRNIHSSDWP KFFEKYLRDV NCPFKIQDRQ EAIDWLLGLA VRLEYGDNAE KYKDLVPDNS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
RTRAF (C14orf166) is an RNA-binding protein involved in modulation of mRNA transcription by Polymerase II (PubMed:16950395). Component of the tRNA-splicing ligase complex and is required for tRNA ligation (PubMed:24870230). May be required for RNA transport (PubMed:24608264).
Specifications
RTRAF | |
Polyclonal | |
Unconjugated | |
RTRAF | |
2700060E02Rik; C14orf166; CGI99; CGI-99; chromosome 14 open reading frame 166; CLE; CLE7; CLE7 homolog; hCLE1; LCRP369; LRRGT00192; RGD1304704; RIKEN cDNA 2700060E02 gene; RLL motif containing 1; RLLM1; RNA transcription, translation and transport factor protein; RTRAF; similar to Hypothetical protein CGI-99; UPF0568 protein C14orf166; UPF0568 protein C14orf166 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y224 | |
RTRAF | |
Recombinant protein corresponding to Human C14orf166. Recombinant protein control fragment (Product #RP-98350). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction