Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTRAF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106393 Supplier Invitrogen™ Supplier No. PA5106393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C14orf166.

RTRAF (C14orf166) is an RNA-binding protein involved in modulation of mRNA transcription by Polymerase II (PubMed:16950395). Component of the tRNA-splicing ligase complex and is required for tRNA ligation (PubMed:24870230). May be required for RNA transport (PubMed:24608264).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTRAF
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RTRAF
Gene Accession No. Q9CQE8, Q9Y224
Gene Alias 2700060E02Rik; C14orf166; CGI99; CGI-99; chromosome 14 open reading frame 166; CLE; CLE7; CLE7 homolog; hCLE1; LCRP369; LRRGT00192; RGD1304704; RIKEN cDNA 2700060E02 gene; RLL motif containing 1; RLLM1; RNA transcription, translation and transport factor protein; RTRAF; similar to Hypothetical protein CGI-99; UPF0568 protein C14orf166; UPF0568 protein C14orf166 homolog
Gene Symbols RTRAF
Host Species Rabbit
Immunogen A synthesized peptide derived from human C14orf166(Accession Q9Y224), corresponding to amino acid residues E24-Q74.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302247, 51637, 68045
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.