Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553720 Supplier Invitrogen™ Supplier No. PA553720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRKEPGARLE ATRGAARPHK QGTKPMITRP SVSQLGEGKC PSSQHLQSLR HNKQHALTLT KARCCGECST CFCTEEKSEC QRHEETSPGS CNHQIMSAST ISAFCATPRF KQLFKGTVEQ MSQM Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 24%.

The function of C14orf177 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf177
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C14orf177
Gene Accession No. Q52M58
Gene Alias C14orf177; chromosome 14 open reading frame 177; putative uncharacterized protein C14orf177
Gene Symbols C14orf177
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C14orf177. Recombinant protein control fragment (Product #RP-88878).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283598
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.