Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf179 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144009
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144009 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144009 Supplier Invitrogen™ Supplier No. PA5144009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human U-87MG whole cell, human T-47D whole cell, human MCF-7 whole cell, human PC-3 whole cell, human A549 whole cell, human Jurkat whole cell, human A431 whole cell, rat testis tissue, rat kidney tissue, rat liver tissue, mouse testis tissue, mouse kidney tissue, mouse liver tissue. ICC/IF: T-47D cell. Flow: Caco-2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

IFT43, encoded by C14ORF179, is a subunit of the intraflagellar transport complex A (IFT-A) of primary cilia. The IFT-A is implicated in retrograde ciliary transport along axonemal microtubules. Defects in subunits of IFT-A has been associated with skeletal ciliopathies, including Sensenbrenner syndrome, Jeune syndrome, and Ellis-van-Creveld syndrome. Mutations in C14ORF179 has been linked to Sensenbrenner syndrome. (21378380).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf179
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene IFT43
Gene Accession No. Q96FT9, Q9DA69
Gene Alias 1700019E19Rik; C14orf179; CED3; IFT complex A subunit; IFT43; intraflagellar transport 43; intraflagellar transport 43 homolog; intraflagellar transport protein 43 homolog; R75064; RGD1307392
Gene Symbols IFT43
Host Species Rabbit
Immunogen E.coli-derived human IFT43 recombinant protein (Position: M1-T208).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112752, 299209, 76411
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.