Learn More
Invitrogen™ C14orf21 Polyclonal Antibody

Description
Immunogen sequence: GGTILESERA RPRGSQSSEA QKTPAQECKP ADFEVPETFL NRLQDLSSSF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 85%; Human: 100%; Mouse: 83%; Rabbit: 92%; Rat: 83%.
Specifications
Specifications
| Antigen | C14orf21 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | NOP9 |
| Gene Accession No. | Q86U38 |
| Gene Alias | C14orf21; KIAA2021; NOP9; NOP9 nucleolar protein; NOP9 nucleolar protein homolog; nucleolar protein 9; pumilio domain-containing protein C14orf21 |
| Gene Symbols | NOP9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.