Learn More
Invitrogen™ C14orf37 Polyclonal Antibody

Description
Immunogen sequence: SQTAQVSHEGM EGGQPWTEAA QVALGLPEGE THTGTALLIA HGNERSPAFT DQSSFTPTSL MEDMKVSIVN LLQSTGDFTE STKENDALFF LETTVSVSVY ESEADQLLGN TMKDIITQEM TTAVQEPDAT LSMVTQEQVA TLELIRDSG Highest antigen sequence identity to the following orthologs - mouse 45%, rat 48%.
Specifications
Specifications
| Antigen | C14orf37 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C14orf37 |
| Gene Accession No. | Q86TY3 |
| Gene Alias | Armadillo-like helical domain-containing protein 4; ARMH4; c14_5376; C14orf37; chromosome 14 open reading frame 37; uncharacterized protein C14orf37; upstream of mTORC2 |
| Gene Symbols | C14orf37 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.