Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551635 Supplier Invitrogen™ Supplier No. PA551635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQTAQVSHEG MEGGQPWTEA AQVALGLPEG ETHTGTALLI AHGNERSPAF TDQSSFTPTS LMEDMKVSIV NLLQSTGDFT ESTKENDALF FLETTVSVSV YESEADQLLG NTMKDIITQE MTTAVQEPDA TLSMVTQEQV ATLELIRDSG Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 48%.

C14orf37 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf37
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C14orf37
Gene Accession No. Q86TY3
Gene Alias Armadillo-like helical domain-containing protein 4; ARMH4; c14_5376; C14orf37; chromosome 14 open reading frame 37; uncharacterized protein C14orf37; upstream of mTORC2
Gene Symbols C14orf37
Host Species Rabbit
Immunogen Recombinant protein fragment of Human C14orf37 (Product #RP-88981)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.