Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100411 Supplier Invitrogen™ Supplier No. PA5100411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C14orf39.

Meiotic protein that localizes to the central element of the synaptonemal complex and is required for chromosome synapsis during meiotic recombination. Required for the appropriate processing of intermediate recombination nodules before crossover formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf39
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C14orf39
Gene Accession No. Q8N1H7, Q9CTN5
Gene Alias 4921504I02Rik; 4930447C04Rik; A930035O15Rik; C14orf39; chromosome 14 open reading frame 39; protein SIX6OS1; RIKEN cDNA 4930447C04 gene; Six6 antisense; Six6 opposite strand transcript 1; Six6 opposite strand transcript 1 homolog; Six6as; Six6OS; Six6os1
Gene Symbols 4930447C04Rik, C14orf39
Host Species Rabbit
Immunogen A synthesized peptide derived from human SIX6OS1(Accession Q8N1H7), corresponding to amino acid residues E528-S578.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317761, 75801
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.