Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563741 Supplier Invitrogen™ Supplier No. PA563741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVDEMEIEIN YLNQQISRHN ETKALSETLE EKNKNTENRK ELKERIFGKD EHVLTLNKTQ SSQLFLPYES QKLVRPIK Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 56%.

Meiotic protein that localizes to the central element of the synaptonemal complex and is required for chromosome synapsis during meiotic recombination. Required for the appropriate processing of intermediate recombination nodules before crossover formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf39
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C14orf39
Gene Accession No. Q8N1H7
Gene Alias 4921504I02Rik; 4930447C04Rik; A930035O15Rik; C14orf39; chromosome 14 open reading frame 39; protein SIX6OS1; RIKEN cDNA 4930447C04 gene; Six6 antisense; Six6 opposite strand transcript 1; Six6 opposite strand transcript 1 homolog; Six6as; Six6OS; Six6os1
Gene Symbols C14orf39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C14orf39. Recombinant protein control fragment (Product #RP-101532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317761
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.